2-Butene-1,4-diol 98% (by GC, sum of isomers)

Supplier: Ambeed
Warning

A830281-100G A830281-25G A830281-500G
76698-242EA 40.32 USD
76698-242 76698-244 76698-246 76698-248
2-Butene-1,4-diol 98% (by GC, sum of isomers)
2-Butene-1,4-diol

Precaución: for Research Use Only

Formula: C₄H₈O₂
MDL Number: MFCD00063207
CAS Number: 110-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR