Dibenzo-30-crown-10 (dibenzo-1,4,7,10,13,16,19,22,25,28-decaoxacyclotriacontane) 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
6,7,9,10,12,13,15,16,23,24,26,27,29,30,32,33-Hexadecahydrodibenzo[b,q][1,4,7,10,13,16,19,22,25,28]decaoxacyclotriacontine, Dibenzo-30-crown-10-ether
A829731-100MG
A829731-1G
A829731-250MG
76698-234EA
45
USD
76698-234
76698-236
76698-238
Dibenzo-30-crown-10 (dibenzo-1,4,7,10,13,16,19,22,25,28-decaoxacyclotriacontane) 98%
Dibenzo-30-crown-10 (dibenzo-1,4,7,10,13,16,19,22,25,28-decaoxacyclotriacontane)
Precaución: for Research Use Only
Formula:
C₂₈H₄₀O₁₀ MW: 536.62 g/mol |
MDL Number:
MFCD00054624 CAS Number: 17455-25-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...