Benzyl-4-hydroxybenzoate 95%

Supplier: Ambeed
Danger

Synonyms: Benzyl 4-hydroxybenzoate, Benzylparaben, 4-Hydroxybenzoic acid benzyl ester, Benzyl Parahydroxybenzoate

A829369-25G A829369-5G
76698-230EA 30 USD
76698-230 76698-232
Benzyl-4-hydroxybenzoate 95%
Benzyl-4-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₁₄H₁₂O₃
MW: 228.25 g/mol
MDL Number: MFCD00016471
CAS Number: 94-18-8
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR