Aminoferrocene 97%

Supplier: Ambeed
Warning

Synonyms: Ferrocenylamine

A828440-100MG A828440-250MG
76698-216EA 39 USD
76698-216 76698-218
Aminoferrocene 97%
Aminoferrocene

Precaución: for Research Use Only

Formula: C₁₀H₂FeN
MW: 191.97 g/mol
MDL Number: MFCD03844760
CAS Number: 1273-82-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR