Cyclohexane-1,4-diylbis(methylene) diacrylate 98%

Supplier: Ambeed
Warning

A827242-1G A827242-250MG
76698-212EA 98.67 USD
76698-212 76698-214
Cyclohexane-1,4-diylbis(methylene) diacrylate 98%
Cyclohexane-1,4-diylbis(methylene) diacrylate

Precaución: for Research Use Only

Formula: C₁₄H₂₀O₄
MW: 252.30 g/mol
MDL Number: MFCD00135698
CAS Number: 67905-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR