Bzl-Ser-oh 98%

Supplier: Ambeed
Warning

Synonyms: N-Bzl-Ser-OH, L-N-Benzylserine

A823793-1G A823793-250MG A823793-25G
76698-192EA 38.2 USD
76698-192 76698-194 76698-196
Bzl-Ser-oh 98%
Bzl-Ser-oh

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₃
MW: 195.218 g/mol
MDL Number: MFCD00020417
CAS Number: 17136-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR