(Formylmethyl)triphenylphosphonium chloride 98%

Supplier: Ambeed
Danger

Synonyms: (2-Oxoethyl)triphenylphosphonium chloride

A822743-25G A822743-5G
76698-188EA 42.44 USD
76698-188 76698-190
(Formylmethyl)triphenylphosphonium chloride 98%
(Formylmethyl)triphenylphosphonium chloride

Precaución: for Research Use Only

Formula: C₂₀H₁₈ClOP
MW: 340.79 g/mol
MDL Number: MFCD00012003
CAS Number: 62942-43-2
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR