N-Boc-trans-4-fluoro-L-proline methyl ester 97%

Supplier: Ambeed
Warning

Synonyms: (2S,4R)-1-tert-Butyl 2-methyl 4-fluoropyrrolidine-1,2-dicarboxylate

A822665-100G A822665-10G A822665-1G A822665-250MG A822665-25G A822665-5G
76698-176EA 795.68 USD
76698-176 76698-178 76698-180 76698-182 76698-184 76698-186
N-Boc-trans-4-fluoro-L-proline methyl ester 97%
N-Boc-trans-4-fluoro-L-proline methyl ester

Precaución: for Research Use Only

Formula: C₁₁H₁₈FNO₄
MW: 247.27 g/mol
MDL Number: MFCD07367993
CAS Number: 203866-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR