8-Bromo-6-methylquinoline 98%

Supplier: Ambeed
Warning

A821530-1G A821530-250MG
76698-172EA 83.82 USD
76698-172 76698-174
8-Bromo-6-methylquinoline 98%
8-Bromo-6-methylquinoline

Precaución: for Research Use Only

Formula: C₁₀H₈BrN
MDL Number: MFCD09701455
CAS Number: 84839-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR