Brilliant green BS

Supplier: Ambeed
Warning

Synonyms: Ethyl Green, CI 42040, Basic Green 1, N-(4-((4-(Diethylamino)phenyl)(phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-ethylethanaminium hydrogen sulphate, Diamond Green, Malachite Green G, Emerald Green, Astradiamant green GX, Solid Green JO, Brilliant Green hydrogen sulphate

A820981-100G A820981-25G A820981-5G A820981-500G
76698-168EA 26 USD
76698-168 76698-170 77603-100 77627-620
Brilliant green BS
Brilliant green

Precaución: for Research Use Only

Formula: C₂₇H₃₄N₂O₄S
MW: 482.65 g/mol
MDL Number: MFCD00011880
CAS Number: 633-03-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR