(Trimethylphosphine)gold(I) chloride 98%

Supplier: Ambeed
Warning

A819934-100MG A819934-1G A819934-250MG
76698-162EA 74 USD
76698-162 76698-164 76698-166
(Trimethylphosphine)gold(I) chloride 98%
(Trimethylphosphine)gold (I) chloride

Precaución: for Research Use Only

Formula: C₃H₉AuClP
MDL Number: MFCD00058861
CAS Number: 15278-97-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR