Magnesium 8-anilinonaphthalene-1-sulphonate hydrate 98%

Supplier: Ambeed
Warning

Synonyms: 8-Anilino-1-naphthalenesulphonic acid magnesium(II) salt, ANS-Mg, 1,8-ANS magnesium salt, N-Phenyl peri acid magnesium salt, ANSA magnesium salt, Magnesium(II) 8-Anilino-1-naphthalenesulphonate Hydrate, Magnesium 8-(phenylamino)naphthalene-1-sulphonate, Magnesium(II) 8-Anilino-1-naphthalenesulphonate, 8-Anilino-1-naphthalenesulphonic acid Magnesium(II) Salt Hydrate, 8-Anilino-1-naphthalenesulphonic acid hemimagnesium salt hydrate

A819084-1G A819084-5G A819084-25G
76698-152EA 6 USD
76698-152 76698-154 77878-668
Magnesium 8-anilinonaphthalene-1-sulphonate hydrate 98%
Magnesium 8-anilinonaphthalene-1-sulphonate hydrate

Precaución: for Research Use Only

Formula: C₃₂H₂₄MgN₂O₆S₂·xH₂O
MDL Number: MFCD00065353
CAS Number: 18108-68-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR