8-Aminooctan-1-ol 98%

Supplier: Ambeed
Warning

Synonyms: 8-Amino-1-octanol

A818879-250MG A818879-5G A818879-1G A818879-25G A818879-100G
76698-150EA 20 USD
76698-150 77552-514 77569-524 77603-086 77627-616
8-Aminooctan-1-ol 98%
8-Aminooctan-1-ol

Precaución: For research use only.

Formula: C₈H₁₉NO
MW: 145.2400 g/mol
MDL Number: MFCD02094315
CAS Number: 19008-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR