4-(Dimethylamino)-2-methoxybenzaldehyde 95%

Supplier: Ambeed
Warning

A814764-1G A814764-250MG A814764-5G
76698-130EA 23 USD
76698-130 76698-132 76698-134
4-(Dimethylamino)-2-methoxybenzaldehyde 95%
4-(Dimethylamino)-2-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MDL Number: MFCD00151814
CAS Number: 84562-48-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR