Chlorhexidine 99+%

Supplier: Ambeed
Danger

Synonyms: 1,1'-Hexamethylenebis[5-(p-chlorophenyl)biguanide]

A814733-25MG A814733-5G A814733-25G A814733-100G A814733-10G A814733-1MG
76698-126EA 5.31 USD
76698-126 76698-128 77595-300 77603-060 77627-606 77627-608
Chlorhexidine 99+%
Chlorhexidine

Precaución: for Research Use Only

Formula: C₂₂H₃₀Cl₂N₁₀
MW: 505.4466 g/mol
MDL Number: MFCD00009673
CAS Number: 55-56-1
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR