8-Methyl-8-azabicyclo[3.2.1]octan-3-amine 95%

Supplier: Ambeed
Danger

Synonyms: 3-Amino-8-methyl-8-azabicyclo[3.2.1]octane

A813975-100MG A813975-1G A813975-250MG
76698-120EA 16.98 USD
76698-120 76698-122 76698-124
8-Methyl-8-azabicyclo[3.2.1]octan-3-amine 95%
8-Methyl-8-azabicyclo[3.2.1]octan-3-amine

Precaución: for Research Use Only

Formula: C₈H₁₆N₂
MW: 140.23 g/mol
MDL Number: MFCD00210699
CAS Number: 98998-25-5
UN: 2733
ADR: 3,8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR