6-Bromo-3-nitropyridin-2-amine 95%

Supplier: Ambeed
Warning

A811389-1G A811389-250MG A811389-5G
76698-114EA 15 USD
76698-114 76698-116 76698-118
6-Bromo-3-nitropyridin-2-amine 95%
6-Bromo-3-nitropyridin-2-amine

Precaución: for Research Use Only

Formula: C₅H₄BrN₃O₂
MDL Number: MFCD07774108
CAS Number: 84487-04-7
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR