4-(Trifluoromethoxy)phenylacetonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 1-(Cyanmethyl)-4-(trifluormethoxy)benzol, 4-(TrifluoroMethoxy)phenylacetonitrile, 4-(Trifluoromethoxy)benzyl cyanide

A811177-100G A811177-1G A811177-25G A811177-5G
76698-106EA 214.31 USD
76698-106 76698-108 76698-110 76698-112
4-(Trifluoromethoxy)phenylacetonitrile 98%
4-(Trifluoromethoxy)phenylacetonitrile

Precaución: for Research Use Only

Formula: C₉H₆F₃NO
MW: 201.15 g/mol
MDL Number: MFCD00061240
CAS Number: 49561-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR