Daphnetin 95%

Supplier: Ambeed
Warning

Synonyms: 7,8-Dihydroxy-2H-chromen-2-one, 7,8-Dihydroxycoumarin

A811058-100MG A811058-1G A811058-1MG A811058-250MG
76698-100EA 36.08 USD
76698-100 76698-102 76698-104 77627-604
Daphnetin 95%
Daphnetin

Precaución: for Research Use Only

Formula: C₉H₆O₄
MW: 178.14 g/mol
MDL Number: MFCD00016977
CAS Number: 486-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR