Sodium 2-((4-(hydroxybis(4-(phenylamino)phenyl)methyl)phenyl)amino)benzenesulfonate indicator

Supplier: Ambeed
Warning

A809503-25G A809503-100G A809503-5G
76698-098EA 19 USD
76698-098 77627-600 77627-602
Sodium 2-((4-(hydroxybis(4-(phenylamino)phenyl)methyl)phenyl)amino)benzenesulfonate indicator
Sodium 2-((4-(hydroxybis(4-(phenylamino)phenyl)methyl)phenyl)amino)benzenesulfonate

Precaución: for Research Use Only

Formula: C₃₇H₃₀N₃NaO₄S
MDL Number: MFCD24369696
CAS Number: 1713165-34-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR