Acetone oxime 97%

Supplier: Ambeed
Danger

Synonyms: Propan-2-one oxime, Acetoxime, Acetone oxime, 2-Propanone oxime

A807939-10G A807939-25G
76698-088EA 5.31 USD
76698-088 76845-326
Acetone oxime 97%
Acetone oxime

Precaución: for Research Use Only

Formula: C₃H₇NO
MW: 73.09 g/mol
MDL Number: MFCD00002118
CAS Number: 127-06-0
UN: 1325
ADR: 4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR