3-(4-(3-Chlorophenyl)piperazin-1-yl)-1,1-diphenylpropan-2-ol dihydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
BRL-15572 2HCl
A807415-10MG
A807415-1MG
A807415-50MG
A807415-5MG
76698-080EA
33.95
USD
76698-080
76698-082
76698-084
76698-086
3-(4-(3-Chlorophenyl)piperazin-1-yl)-1,1-diphenylpropan-2-ol dihydrochloride 98%
3-(4-(3-Chlorophenyl)piperazin-1-yl)-1,1-diphenylpropan-2-ol dihydrochloride
Precaución: for Research Use Only
Formula:
C₂₅H₂₉Cl₃N₂O MW: 479.87 g/mol |
MDL Number:
MFCD02684402 CAS Number: 193611-72-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...