β-Caryophyllene 90%

Supplier: Ambeed
Warning

A805906-100G A805906-25G A805906-500G A805906-5G A805906-1000G
76698-070EA 20 USD
76698-070 76698-072 76698-074 76698-076 77878-650
β-Caryophyllene 90%
β-Caryophyllene

Precaución: for Research Use Only

Formula: C₁₅H₂₄
MDL Number: MFCD00075925
CAS Number: 87-44-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR