7,8-Dihydroxy-6-methoxy-2H-chromen-2-one 95%

Supplier: Ambeed
Warning

Synonyms: Fraxetin

A805736-1MG A805736-250MG A805736-1G
76698-068EA 5.31 USD
76698-068 77603-044 77627-596
7,8-Dihydroxy-6-methoxy-2H-chromen-2-one 95%
7,8-Dihydroxy-6-methoxy-2H-chromen-2-one

Precaución: for Research Use Only

Formula: C₁₀H₈O₅
MW: 208.16 g/mol
MDL Number: MFCD00006873
CAS Number: 574-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR