Methyl-4,5-dimethoxy-3-hydroxybenzoate 95%

Supplier: Ambeed
Warning

A804691-1G A804691-5G A804691-250MG
76698-062EA 70 USD
76698-062 76698-064 77627-594
Methyl-4,5-dimethoxy-3-hydroxybenzoate 95%
Methyl-4,5-dimethoxy-3-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₅
MDL Number: MFCD00598192
CAS Number: 83011-43-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR