7-Fluoro-3,4-dihydro-2H-1,4-benzoxazine 97%

Supplier: Ambeed
Warning

Synonyms: 7-Fluoro-3,4-dihydro-2H-benzo[b][1,4]oxazine

A801727-100MG A801727-1G A801727-250MG A801727-5G
76698-042EA 43.5 USD
76698-042 76698-044 76698-046 76698-048
7-Fluoro-3,4-dihydro-2H-1,4-benzoxazine 97%
7-Fluoro-3,4-dihydro-2H-1,4-benzoxazine

Precaución: for Research Use Only

Formula: C₈H₈FNO
MW: 153.16 g/mol
MDL Number: MFCD11848177
CAS Number: 56346-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR