cis-1,2-Cyclohexanedimethanol 95%

Supplier: Ambeed
Warning

A800637-250MG A800637-1G A800637-5G
76698-040EA 15 USD
76698-040 77880-594 77880-595
cis-1,2-Cyclohexanedimethanol 95%
cis-1,2-Cyclohexanedimethanol

Precaución: for Research Use Only

Formula: C₈H₁₆O₂
MDL Number: MFCD00062987
CAS Number: 15753-50-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR