cis-Stilbene 96%

Supplier: Ambeed
Warning

Synonyms: cis-1,2-Diphenylethene, Isostilbene, cis-1,2-Diphenylethylene, (Z)-1,2-Diphenylethene

A799290-100G A799290-1G A799290-5G
76698-034EA 250 USD
76698-034 76698-036 76698-038
cis-Stilbene 96%
cis-Stilbene

Precaución: for Research Use Only

Formula: (C₆H₅)CHCH(C₆H₅)
MW: 180.25 g/mol
Boiling Pt: 307
MDL Number: MFCD00004788
CAS Number: 645-49-8
Merck Index: 12,08972

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR