(S)-N-Methyl-N-methoxy-2-(tert-butoxycarbonylamino)-4-methylpentanamide 98%

Supplier: Ambeed
Warning

Synonyms: Boc-Leu-NMe(OMe)

A799251-10G A799251-1G A799251-25G
76698-028EA 67.9 USD
76698-028 76698-030 76698-032
(S)-N-Methyl-N-methoxy-2-(tert-butoxycarbonylamino)-4-methylpentanamide 98%
(S)-N-Methyl-N-methoxy-2-(tert-butoxycarbonylamino)-4-methylpentanamide

Precaución: for Research Use Only

Formula: C₁₃H₂₆N₂O₄
MW: 274.35 g/mol
MDL Number: MFCD00151875
CAS Number: 87694-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR