4-Iodocumene 95%

Supplier: Ambeed
Warning

Synonyms: 4-Iodoisopropylbenzene, 1-Iodo-4-isopropylbenzene, 2-(4-Iodophenyl) propane

A798482-100G A798482-25G A798482-500G A798482-5G
76698-020EA 268.41 USD
76698-020 76698-022 76698-024 76698-026
4-Iodocumene 95%
4-Iodocumene

Precaución: for Research Use Only

Formula: C₉H₁₁I
MW: 246.09 g/mol
MDL Number: MFCD00013711
CAS Number: 17356-09-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR