Bromocresol purple indicator
Supplier: Ambeed
|
Warning
|
Synonyms:
5',5''-Dibromo-o-cresolsulphonphthalein, 3,3-Bis(3-bromo-4-hydroxy-5-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
A795101-100G
A795101-25G
76697-984EA
63
USD
76697-984
76697-986
Bromocresol purple indicator
Bromocresol purple
Precaución: for Research Use Only
Formula:
C₂₁H₁₆Br₂O₅S MW: 540.23 g/mol Melting Pt: 242 |
MDL Number:
MFCD00011681 CAS Number: 115-40-2 Merck Index: 13,01371 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...