Dimethyl-bis-(phenylethynyl)silane 98%

Supplier: Ambeed
Warning

A793209-1G A793209-25G A793209-5G
76697-976EA 16 USD
76697-976 76697-978 76697-980
Dimethyl-bis-(phenylethynyl)silane 98%
Dimethyl-bis-(phenylethynyl)silane

Precaución: for Research Use Only

Formula: C₁₈H₁₆Si
MDL Number: MFCD00270967
CAS Number: 2170-08-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR