Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')zinc(II) 98%

Supplier: Ambeed
Warning

Synonyms: Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)zinc(II), Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)zinc, Zn(TMHD)2

A790151-1G A790151-250MG A790151-5G
76697-964EA 40.32 USD
76697-964 76697-966 76697-968
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')zinc(II) 98%
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')zinc(II)

Precaución: for Research Use Only

Formula: C₂₂H₃₈O₄Zn
MW: 431.92 g/mol
MDL Number: MFCD00192578
CAS Number: 14363-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR