(-)-3-(3,4-Dihydroxyphenyl)-2-methyl-L-alanine sesquihydrate 98+%

Supplier: Ambeed
Warning

Synonyms: (S)-2-amino-3-(3,4-dihydroxyphenyl)-2-methylpropanoic acid sesquihydrate, Methyldopa sesquihydrate

A789550-5G A789550-100G A789550-25G
76697-960EA 7 USD
76697-960 77880-570 77880-571
(-)-3-(3,4-Dihydroxyphenyl)-2-methyl-L-alanine sesquihydrate 98+%
(-)-3-(3,4-Dihydroxyphenyl)-2-methyl-L-alanine sesquihydrate

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₄·1.5H₂O
MW: 238.24 g/mol (211.22 anhy)
MDL Number: MFCD00149280
CAS Number: 41372-08-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR