2H-1,3-Benzodioxol-4-ylmethanamine 95%

Supplier: Ambeed
Warning

Synonyms: 1,3-Benzodioxole-4-methanamine

A789444-1G A789444-250MG
76697-956EA 311.91 USD
76697-956 76697-958
2H-1,3-Benzodioxol-4-ylmethanamine 95%
2H-1,3-Benzodioxol-4-ylmethanamine

Precaución: for Research Use Only

Formula: C₈H₉NO₂
MW: 151.163 g/mol
MDL Number: MFCD11858181
CAS Number: 182634-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR