Glycidyl-4-methoxyphenyl ether 98%

Supplier: Ambeed
Warning

Synonyms: 2-((4-Methoxyphenoxy)methyl)oxirane

A785021-1G A785021-5G A785021-25G
76697-936EA 9 USD
76697-936 76697-938 77880-564
Glycidyl-4-methoxyphenyl ether 98%
Glycidyl-4-methoxyphenyl ether

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MDL Number: MFCD00005135
CAS Number: 2211-94-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR