2-[Bis(2-hydroxyethyl)amino]ethanesulfonic acid sodium salt 99%

Supplier: Ambeed
Warning

A784302-100G A784302-25G A784302-500G
76697-922EA 41 USD
76697-922 76697-924 76697-926
2-[Bis(2-hydroxyethyl)amino]ethanesulfonic acid sodium salt 99%
2-[Bis(2-hydroxyethyl)amino]ethanesulphonic acid sodium salt

Precaución: for Research Use Only

Formula: C₆H₁₄NNaO₅S
MDL Number: MFCD00067425
CAS Number: 66992-27-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR