5-(3,4-Dimethoxybenzyl)pyrimidine-2,4-diamine 98%

Supplier: Ambeed
Warning

A783370-100G A783370-1G A783370-1MG A783370-250MG A783370-25G A783370-5G
76697-906EA 781 USD
76697-906 76697-908 76697-910 76697-912 76697-914 76697-916
5-(3,4-Dimethoxybenzyl)pyrimidine-2,4-diamine 98%
5-(3,4-Dimethoxybenzyl)pyrimidine-2,4-diamine

Precaución: for Research Use Only

Formula: C₁₃H₁₆N₄O₂
MDL Number: MFCD00057349
CAS Number: 5355-16-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR