(±)-δ-Hexalactone 98% (by GC)

Supplier: Ambeed
Warning

Synonyms: (±)-Hexan-5-olide, (±)-Tetrahydro-6-methyl-2H-pyran-2-one, 6-Methyltetrahydro-2H-pyran-2-one, (±)-δ-Caprolactone

A782970-100G A782970-25G A782970-5G
76697-900EA 137.92 USD
76697-900 76697-902 76697-904
(±)-δ-Hexalactone 98% (by GC)
(±)-δ-Hexalactone

Precaución: for Research Use Only

Formula: C₆H₁₀O₂
MW: 114.14 g/mol
MDL Number: MFCD00083574
CAS Number: 823-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR