1-(4-Chlorophenyl)-2-(4-(4-fluorobenzyl)piperidin-1-yl)ethan-1-ol 99+%
Supplier: Ambeed
|
Warning
|
A782266-100MG
A782266-10MG
A782266-1MG
A782266-250MG
A782266-25MG
A782266-50MG
A782266-5MG
76697-884EA
498.63
USD
76697-884
76697-886
76697-888
76697-890
76697-892
76697-894
76697-896
1-(4-Chlorophenyl)-2-(4-(4-fluorobenzyl)piperidin-1-yl)ethan-1-ol 99+%
1-(4-Chlorophenyl)-2-(4-(4-fluorobenzyl)piperidin-1-yl)ethan-1-ol
Precaución: for Research Use Only
Formula:
C₂₀H₂₃ClFNO MW: 347.85 g/mol |
MDL Number:
MFCD00866651 CAS Number: 119431-25-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...