Diazolidinyl urea 98%

Supplier: Ambeed
Warning

Synonyms: 1,3-Bis(hydroxymethyl)-5-[1,3-bis(hydroxymethyl)ureido]hydantoin, 1-(1,3-Bis(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl)-1,3-bis(hydroxymethyl)urea

A781898-100G A781898-10G A781898-25G A781898-500G A781898-1000G
76697-876EA 29.71 USD
76697-876 76697-878 76697-880 76697-882 77627-558
Diazolidinyl urea 98%
Diazolidinyl urea

Precaución: for Research Use Only

Formula: C₈H₁₄N₄O₇
MW: 278.22 g/mol
MDL Number: MFCD03547942
CAS Number: 78491-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR