1H-Imidazole-4-propanamide, α(acetylamino)-N-methyl-(S)- 97%

Supplier: Ambeed
Warning

Synonyms: (S)-2-Acetamido-3-(1H-imidazol-4-yl)-N-methylpropanamide, Ac-His-NHMe

A779177-1G A779177-250MG
76697-866EA 200.52 USD
76697-866 76697-868
1H-Imidazole-4-propanamide, α(acetylamino)-N-methyl-(S)- 97%
1H-Imidazole-4-propanamide, alpha(acetylamino)-N-methyl-(S)-

Precaución: for Research Use Only

Formula: C₉H₁₄N₄O₂
MW: 210.237 g/mol
MDL Number: MFCD00237489
CAS Number: 6367-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR