cis-1,2-Bis(diphenylphosphino)ethylene 98%

Supplier: Ambeed
Warning

Synonyms: (Z)-1,2-Bis(diphenylphosphino)ethene

A774134-100MG A774134-1G A774134-250MG A774134-5G
76697-836EA 17 USD
76697-836 76697-838 76697-840 76697-842
cis-1,2-Bis(diphenylphosphino)ethylene 98%
cis-1,2-Bis(diphenylphosphino)ethylene

Precaución: for Research Use Only

Formula: C₂₆H₂₂P₂
MDL Number: MFCD00003046
CAS Number: 983-80-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR