7-(Diethylamino)-3-(1-methyl-2-benzimidazolyl)coumarin 98%

Supplier: Ambeed
Warning

Synonyms: Coumarin 30, 3-(2-N-Methylbenzimidazolyl)-7-N,N-diethylaminocoumarin, 7-(Diethylamino)-3-(1-methyl-1H-benzo[d]imidazol-2-yl)-2H-chromen-2-one

A771926-100MG A771926-250MG
76697-828EA 40.32 USD
76697-828 76697-830
7-(Diethylamino)-3-(1-methyl-2-benzimidazolyl)coumarin 98%
7-(Diethylamino)-3-(1-methyl-2-benzimidazolyl)coumarin

Precaución: for Research Use Only

Formula: C₂₁H₂₁N₃O₂
MW: 347.42 g/mol
MDL Number: MFCD00051349
CAS Number: 41044-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR