(R)-4,5-Diamino-5-oxopentanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: H-D-Glu-NH₂

A771454-1G A771454-250MG A771454-5G
76697-822EA 149.59 USD
76697-822 76697-824 76697-826
(R)-4,5-Diamino-5-oxopentanoic acid 98%
(R)-4,5-Diamino-5-oxopentanoic acid

Precaución: for Research Use Only

Formula: C₅H₁₀N₂O₃
MW: 146.146 g/mol
MDL Number: MFCD00237403
CAS Number: 19522-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR