Ethyl-2-cyclohexylidenacetate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-cyclohexylideneacetate

A768664-1G A768664-5G A768664-25G
76697-806EA 20 USD
76697-806 76697-808 77627-524
Ethyl-2-cyclohexylidenacetate 97%
Ethyl-2-cyclohexylidenacetate

Precaución: for Research Use Only

Formula: C₁₀H₁₆O₂
MDL Number: MFCD00963857
CAS Number: 1552-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR