3-(Aminomethyl)benzo[b]thiophene 97%

Supplier: Ambeed
Warning

A768589-100MG A768589-250MG A768589-1G A768589-5G
76697-804EA 16 USD
76697-804 77602-964 77878-613 77878-614
3-(Aminomethyl)benzo[b]thiophene 97%
3-(Aminomethyl)benzo[b]thiophene

Precaución: for Research Use Only

Formula: C₉H₉NS
MDL Number: MFCD00176440
CAS Number: 40615-04-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR