Benorilate 98%

Supplier: Ambeed
Warning

Synonyms: Aspirin Acetaminophen ester, 4-Acetamidophenyl 2-Acetoxybenzoate

A768174-100MG A768174-10G A768174-1G A768174-5G
76697-796EA 5.31 USD
76697-796 76697-798 76697-800 77627-522
Benorilate 98%
Benorilate

Precaución: for Research Use Only

Formula: C₁₇H₁₅NO₅
MW: 313.31 g/mol
MDL Number: MFCD00864257
CAS Number: 5003-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR