α-Methyl-D-phenylalanine 98%

Supplier: Ambeed
Warning

A765142-1G A765142-250MG A765142-100MG
76697-778EA 112 USD
76697-778 76697-780 77880-541
α-Methyl-D-phenylalanine 98%
α-Methyl-D-phenylalanine

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MDL Number: MFCD00153491
CAS Number: 17350-84-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR