9,9-Bis(4-amino-3-methylphenyl)fluorene 95%

Supplier: Ambeed
Warning

Synonyms: 4,4'-(9H-Fluorene-9,9-diyl)bis(2-methylaniline), 2,2'-Dimethyl-4,4'-(9-fluorenylidene)dianiline, 4,4'-(9-Fluorenylidene)di-o-toluidine

A761986-100G A761986-25G A761986-5G
76697-766EA 21 USD
76697-766 76697-768 77627-514
9,9-Bis(4-amino-3-methylphenyl)fluorene 95%
9,9-Bis(4-amino-3-methylphenyl)fluorene

Precaución: for Research Use Only

Formula: C₂₇H₂₄N₂
MW: 376.50 g/mol
MDL Number: MFCD04038168
CAS Number: 107934-60-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR